Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0546110
GREENER_CHOICESDP

Catalog No. 795650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-565-0 50 mg
79-561-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 79-565-0 Supplier Tocris Bioscience™ Supplier No. 7956/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective SLO3 potassium channel inhibitor

VU 0546110 is a selective inhibitor of the sperm-specific potassium channel SLO3 (IC50 = 64 μM). It shows 40-fold selectivity over SLO1. It completely blocks heterologous SLO3 currents and endogenous K+ currents. VU 0546110 inhibits sperm hyperpolarisation and prevents its hyperactivated mobility.

Specifications

Chemical Name or Material 1-[4-[[(4-Methoxyphenyl)thio]methyl]-1-piperidinyl]-2-[4-[(1-methylethyl)sulfonyl]phenyl]ethanone
CAS 1421523-00-3
Quantity 50 mg
Target Non-selective/Other Potassium Channel Blockers
Molecular Formula C24H31NO4S2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.