Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0650991
GREENER_CHOICE

Catalog No. 635510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-551-0 10 mg
63-555-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-551-0 Supplier Tocris Bioscience™ Supplier No. 6355/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-competitive GLP-1 antagonist; orally bioavailable and brain penetrant

VU 0650991 is a noncompetitive glucagon-like peptide-1 (GLP-1) receptor antagonist (IC50 = 0.65 - 0.69 μM). Displays >40-fold selectivity for GLP-1 receptor over glucagon receptors. Blocks activity of Exendin-4 (Cat. No. 1933) in potentiating insulin secretion from primary mouse pancreatic islets in vitro. Decreases blood insulin and increases blood glucose levels in vivo. Brain penetrant and orally bioavailable.

Specifications

Quantity 10 mg
Molecular Formula C16H11ClF6N4O2
Formula Weight 440.73
Product Type Antagonist
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.