Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 152100
GREENER_CHOICE

Catalog No. 338310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-831-0 10 mg
33-835-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-831-0 Supplier Tocris Bioscience™ Supplier No. 3383/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of M4 receptors

Selective positive allosteric modulator of M4 muscarinic acetylcholine receptors (mAChRs) (EC50 = 380 nM). Induces 21-fold shift in ACh potency at M4 receptor. Displays no activity at other mAChR subtypes.

Chemical Identifiers

CAS 409351-28-6
Molecular Formula C18H19N3O2S

Specifications

Quantity 10 mg
Formula Weight 341.43
Purity >98%
Molecular Weight (g/mol) 341.429
Chemical Name or Material VU 152100
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.