Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 6028418 hydrochloride
GREENER_CHOICE

Catalog No. 765850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-585-0 50 mg
76-581-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-585-0 Supplier Tocris Bioscience™ Supplier No. 7658/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective M4 mAChR antagonist

VU 6028418 hydrochloride is a potent and selective M4 mAChR antagonist (IC50 = 4.1 nM and 57 nM for human and rat M4 respectively). VU 6028418 hydrochloride dose-dependently reduces the latency to withdraw and reverses cataleptic behavior in haloperidol -treated animals. Orally bioavailable.

Specifications

Chemical Name or Material (3 aR,5 s,6 AS)-2-((Tetrahydro-2 H-pyran-4-yl)methyl)-N-(6-(2,3,5-trifluorophenyl)pyridazin-3-yl)octahydrocyclopenta[c]pyrrol-5-amine hydrochloride
Quantity 50 mg
Target mAChR M4 Antagonists
Molecular Formula C23H27F3N4O.HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.