Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streamlight™ Vulcan 180 Haz-Lo Class 1 Div 1 Intrinsically Safe Rechargeable Lantern
Shop All Streamlight Products
Click to view available options
Color:
Orange
Yellow
Includes:
Quick Release Buckle Strap, 12 V DC Vehicle-mountable Direct Wire Charging Rack
Quick Release Buckle Strap, Charging Rack, AC/ DC Chargers
Quick Release Shoulder Strap, Rechargeable Battery
Description
- Developed for use in hazardous locations on a portable scene light platform the Class 1, Div. 1 safety rated Vulcan 180 is lightweight and rechargeable
- Combine the three extreme bright white LEDs with cordless operation to eliminate trip hazards while two ultra-bright blue taillights provide visibility in thick smoke
- Features an articulating head that aims the beam where you need it; head locks in-place in forward facing position
- Light Output: High for a bright, far-reaching beam: 400 lumens; 316m beam; runs 10 hours, Low for a less intense beam and longer run time: 130 lumens; 128m beam; runs 24 hours
- Three LEDs produce a tight beam with optimum peripheral illumination
- Two ultra-bright blue taillight LEDs make certain you can be seen even in thick smoke; can be programmed on/off or flashing
- Optimized electronics provide regulated intensity
- Large, multi-function, push-button switch easy to use with gloved hands
- Rechargeable lithium Ion battery. Fully recharges in 15 hours with battery status indicator
Specifications
Specifications
| Light Source | LED |
| Lighting Life | 10 hr. (High), 24 hr. (Low) |
| Voltage | 3.7 V |
| Battery Type | Lithium-ion |
| Certifications/Compliance | CLASS I, II, DIVISION 1, CLASS I, ZONE 0, GROUP IICGRPS A,B,C,D,E,F,G CLASS III; T-CODE: T4Meets applicable European Community Directives.Meets requirements of NFPA 1901-14.1.10.2 (2016) mounted in any position.CEC Compliant. |
| Color | Yellow |
| Includes | Quick Release Buckle Strap, 12 V DC Vehicle-mountable Direct Wire Charging Rack |
| Length (English) | 7.43 in. |
| Material | Nylon |
| Product Type | Lantern/Scene Lighting |
CLASS I, II, DIVISION 1, CLASS I, ZONE 0, GROUP IICGRPS A,B,C,D,E,F,G CLASS III; T-CODE: T4Meets applicable European Community Directives.Meets requirements of NFPA 1901-14.1.10.2 (2016) mounted in any position.CEC Compliant.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction