Learn More
Description
In vivo, VVD 844 slows growth of RAS mutant tumors and human epidermal growth factor receptor 2 (HER2)-overexpressing tumors, without causing hyperglycemia. Exhibits synergistic effects with Sotorasib (Cat. No. 7713) and Adagrasib (Cat. No. 7488). VVD 844 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 5-((1R,2S,5S)-2-(2-Chloro-3-fluorophenyl)-3-azabicyclo[3.1.0]hexan-3-yl)-N-((R,E)-4-(methylsulfonyl)but-3-en-2-yl)pyrazine-2-carboxamide |
| CAS | 3080226-63-4 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | GTPase Inhibitors |
| Molecular Formula | C21H22ClFN4O3S |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
