Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121505
This item is not returnable.
View return policy
Description
VWA1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
VWA1 (also known as WARP) is a member of the von Willebrand factor A-domain (VA-domain) superfamily of extracellular matrix proteins which may play a role in cartilage structure and function. VWA1 is a multimeric component of the chondrocyte pericellular matrix in articular cartilage and intervertebral disc, where it interacts with the basement membrane heparan sulfate proteoglycan perlecan. VWA1 may have a role in maintaining the blood-brain barrier.
Specifications
VWA1 | |
Polyclonal | |
Unconjugated | |
Vwa1 | |
4932416A11Rik; RGD1311476; von Willebrand factor A domain containing 1; von Willebrand factor A domain-containing protein 1; von Willebrand factor A domain-related protein; Vwa1; Warp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
246228 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q8R2Z5 | |
Vwa1 | |
Recombinant protein Von Willebrand Factor A Domain Containing Protein 1. The antigen corresponds to amino acid range 135-366 of the target protein. | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction