Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VWA3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559839 Supplier Invitrogen™ Supplier No. PA559839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KCCDSFNLLS FAESLQSWQD TLVETTDAAC HEAMQWVTHL QAQGSTSILQ ALLKAFSFHD LEGLYLLTDG KPDTSCSLVL NEVQKLREKR DVKVHTISLN CSDRAAVEFL RKLA Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen VWA3A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VWA3A
Gene Accession No. A6NCI4
Gene Alias von Willebrand factor A domain containing 3A; von Willebrand factor A domain-containing protein 3A; VWA3A
Gene Symbols VWA3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VWA3A. Recombinant protein control fragment (Product #RP-98773).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146177
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.