Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWA3A Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $482.00
Specifications
Antigen | VWA3A |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119506
|
Invitrogen™
PA5119506 |
200 μL |
Each for $482.00
|
|
|||||
PIPA5121503
|
Invitrogen™
PA5121503 |
20 μL |
Each for $178.40
|
|
|||||
Description
VWA3A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
VWA3A | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
VWA3A | |
von Willebrand factor A domain containing 3A; von Willebrand factor A domain-containing protein 3A; VWA3A | |
VWA3A | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
A6NCI4 | |
146177 | |
Recombinant protein Von Willebrand Factor A Domain Containing Protein 3A. The antigen corresponds to amino acid range 981-1176 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title