Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VWA3B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | VWA3B |
---|---|
Dilution | Western Blot 1:100 - 1:250, Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
VWA3B Polyclonal specifically detects VWA3B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VWA3B | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ32686, MGC26733, von Willebrand factor A domain containing 3B, von Willebrand factor A domain-containing protein 3B | |
VWA3B | |
IgG | |
Affinity Purified |
Western Blot 1:100 - 1:250, Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Polyclonal | |
Rabbit | |
Human | |
Q502W6 | |
200403 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LTGGEFHFYNFGCKDPTPPEAVQNEDLTLLVKEMEQGHSDLEKMQDLYSESLIMDWWYNAEKDGDSKHQKEICSMISTPEKCAKP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title