Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWA5A Monoclonal Antibody (OTI3E6), TrueMAB™, OriGene
SDP

Catalog No. 502542180
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2180 30 μL
50-167-2706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2180 Supplier OriGene Supplier No. TA501570S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VWA5A Monoclonal antibody specifically detects VWA5A in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen VWA5A
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI3E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Vwa5a
Gene Accession No. O00534
Gene Alias 5830475I06Rik; AW552491; BCSC1; BCSC-1; breast cancer suppressor candidate 1; E130119J22; Loh11cr2a; loss of heterozygosity 11 chromosomal region 2 gene A protein; Loss of heterozygosity 11 chromosomal region 2 gene A protein homolog; loss of heterozygosity, 11, chromosomal region 2, gene A; loss of heterozygosity, 11, chromosomal region 2, gene A homolog; Masa1; masa-1; Mast cell surface antigen 1; mast cell surface antigen-1; ortholog of mouse AW551984; von Willebrand factor A domain containing 5A; von Willebrand factor A domain-containing protein 5A; VWA5A
Host Species Mouse
Immunogen Full length human recombinant protein of human VWA5A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4013
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.