Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VWA5B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566723 Supplier Invitrogen™ Supplier No. PA566723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CNIISKYTAFV PVDVSKSRYL PTVVEYPNSG AALRMLGSRA LAQQWRGTSS GFGRPQTMLG EDSAPGN Highest antigen sequence identity to the following orthologs - mouse 62%, rat 57%.

VWA5B1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VWA5B1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VWA5B1
Gene Accession No. Q5TIE3
Gene Alias von Willebrand factor A domain containing 5B1; von Willebrand factor A domain-containing protein 5B1; VWA5B1
Gene Symbols VWA5B1
Host Species Rabbit
Immunogen Recombinant Human VWA5B1. Recombinant protein control fragment (Product #RP-107376).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.