Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWA5B2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563955
Description
Immunogen sequence: LPTVVYSKGL QRGSPAGAWD SDQNGNSKRA LGDPATPTEG PRRPPPRPPC RLSMGRRHKL CSPDPGQANN SEGSDHDYL Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 52%.

Specifications
VWA5B2 | |
Polyclonal | |
Unconjugated | |
VWA5B2 | |
von Willebrand factor A domain containing 5B2; von Willebrand factor A domain-containing protein 5B2; VWA5B2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90113 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N398 | |
VWA5B2 | |
Recombinant protein corresponding to Human VWA5B2. Recombinant protein control fragment (Product #RP-103401). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction