Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VWA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559123 Supplier Invitrogen™ Supplier No. PA559123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTADVQVFPR PEPFVVDEEI DPIPKVINTD LEIVGFIDIA DISSPPVLSR HLVLPIALNK EGDEVGTGIT DDNEDENSAN QIAGKIPNFC VLLHGSLKVE GMVAIVQL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

C15orf44 is an uncharacterized protein that bears a P-loop nucleoside triphosphate hydrolase (NTPase) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VWA9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INTS14
Gene Accession No. Q66H58, Q8R3P6, Q96SY0
Gene Alias 2010321M09Rik; C15orf44; Integrator complex subunit 14; Ints14; RGD1359616; UPF0464 protein C15orf44; UPF0464 protein C15orf44 homolog; von Willebrand factor A domain containing 9; von Willebrand factor A domain-containing protein 9; VWA9
Gene Symbols INTS14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VWA9. Recombinant protein control fragment (Product #RP-98074).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300782, 69882, 81556
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.