Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VWA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113242
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113242 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113242 Supplier Invitrogen™ Supplier No. PA5113242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VWA9 Polyclonal Antibody for Western Blot, IHC (P), ELISA

C15orf44 is an uncharacterized protein that bears a P-loop nucleoside triphosphate hydrolase (NTPase) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VWA9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene INTS14
Gene Accession No. Q8R3P6, Q96SY0
Gene Alias 2010321M09Rik; C15orf44; Integrator complex subunit 14; Ints14; RGD1359616; UPF0464 protein C15orf44; UPF0464 protein C15orf44 homolog; von Willebrand factor A domain containing 9; von Willebrand factor A domain-containing protein 9; VWA9
Gene Symbols INTS14
Host Species Rabbit
Immunogen Recombinant Human Integrator complex subunit 14 protein (231-518AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69882, 81556
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.