Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWC2 Monoclonal Antibody (10A5B4)
GREENER_CHOICE

Catalog No. PIMA531010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531010 Supplier Thermo Scientific Supplier No. MA531010
Add to Cart
Add to Cart

VWC2 Monoclonal Antibody (10A5B4)

VWC2 Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen VWC2
Applications ELISA
Classification Monoclonal
Clone 10A5B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Vwc2
Gene Accession No. Q2TAL6
Gene Alias A930041G11Rik; brain-specific chordin-like protein; Brorin; CR (chordin-like cysteine-rich) domain-containing adhesive protein; Cradin; G11; PSST739; UNQ739; UNQ739/PRO1434; von Willebrand factor C domain containing 2; von Willebrand factor C domain-containing protein 2; Vwc2
Gene Symbols Vwc2
Host Species Mouse
Immunogen Recombinant Human VWC2 Protein (Met1-Met325).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.