Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563097
Description
Immunogen sequence: KLKQAWVSQG GGAKAGDLQV RPRGDTPQAE ALAAAAQDAI GPELAPTPEP PEEYVYPDYR Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.

Specifications
VWC2 | |
Polyclonal | |
Unconjugated | |
Vwc2 | |
A930041G11Rik; brain-specific chordin-like protein; Brorin; CR (chordin-like cysteine-rich) domain-containing adhesive protein; Cradin; G11; PSST739; UNQ739; UNQ739/PRO1434; von Willebrand factor C domain containing 2; von Willebrand factor C domain-containing protein 2; Vwc2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2TAL6 | |
Vwc2 | |
Recombinant protein corresponding to Human VWC2. Recombinant protein control fragment (Product #RP-101583). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction