Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWCE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550896
Description
The antibody detects endogenous levels of total VWCE protein.
May be a regulatory element in the beta-catenin signaling pathway and a target for chemoprevention of hapatocellular carcinoma.
Specifications
VWCE | |
Polyclonal | |
Unconjugated | |
VWCE | |
1300015B04Rik; HBV X protein up-regulated gene 11 protein; HBxAg up-regulated gene 11 protein; RGD1309910; URG11; von Willebrand factor C and EGF domain-containing protein; von Willebrand factor C and EGF domains; VWC1; Vwce | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220001 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96DN2 | |
VWCE | |
Synthetic peptide of human von Willebrand factor C and EGF domains. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction