Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWCE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559160
Description
Immunogen sequence: EKVRCEAACS HPIPSRDGGC CPSCTGCFHS GVVRAEGDVF SPPNENCTVC VCLAGNVSCI SPECPSGPCQ TPPQTDCCTC VPVRCYFHG Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
May be a regulatory element in the beta-catenin signaling pathway and a target for chemoprevention of hapatocellular carcinoma.
Specifications
VWCE | |
Polyclonal | |
Unconjugated | |
VWCE | |
1300015B04Rik; HBV X protein up-regulated gene 11 protein; HBxAg up-regulated gene 11 protein; RGD1309910; URG11; von Willebrand factor C and EGF domain-containing protein; von Willebrand factor C and EGF domains; VWC1; Vwce | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
220001 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DN2 | |
VWCE | |
Recombinant protein corresponding to Human VWCE. Recombinant protein control fragment (Product #RP-97457). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction