Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWCE Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984555
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4555 Supplier Bioss Supplier No. BS2169R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be a regulatory element in the beta-catenin signaling pathway and a target for chemoprevention of hapatocellular carcinoma.

Specifications

Antigen VWCE
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VWCE
Gene Accession No. Q96DN2
Gene Alias HBV X protein up-regulated gene 11 protein; HBxAg up-regulated gene 11 protein; URG11; von Willebrand factor C and EGF domain-containing protein; von Willebrand factor C and EGF domains; VWC1; VWCE
Gene Symbols VWCE
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VWCE.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220001
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.