Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWDE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144764
Description
Immunogen sequence: LLTTQQITVR LNDDCDAETR VTIEVTVKSC DCLNGGSCVS DRNFSPGSGV YLCVCLPGFH GSLCEVDISG CQSNPCGLGS YISGFHSYSC DCPPELKVET QFVNQFTTQT VVLTRSDKSV N Antigen sequence identity: Mouse 56%; Rat 28%.

Specifications
VWDE | |
Polyclonal | |
Unconjugated | |
VWDE | |
von Willebrand factor D and EGF domain-containing protein; von Willebrand factor D and EGF domains; VWDE | |
Rabbit | |
Affinity chromatography | |
RUO | |
221806 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N2E2 | |
VWDE | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1152-1272. Recombinant protein control fragment (Product #RP-109741). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction