Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWDE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555358
Description
Immunogen sequence: YRSVRFDSWH LQQSAVQDLI CDHSLSPGWY RFLILDRPAE MPTKCVEMNH CGTQAPIWLS LRDSETLPSP GEIKQLTACA TWQFLFSTTK DCCLFQIPVS VRNCGNFFVY LLQPTQGCMG YCA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

Specifications
VWDE | |
Polyclonal | |
Unconjugated | |
VWDE | |
von Willebrand factor D and EGF domain-containing protein; von Willebrand factor D and EGF domains; VWDE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221806 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N2E2 | |
VWDE | |
Recombinant protein corresponding to Human VWDE. Recombinant protein control fragment (Product #RP-88710). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction