Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
vWF-A2 Antibody (7G2B11C10), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306526C
Description
vWF-A2 Monoclonal antibody specifically detects vWF-A2 in Human samples. It is validated for ELISASpecifications
vWF-A2 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
7450 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
7G2B11C10 | |
ELISA | |
F8VWF, VWD, VWF von Willebrand factor | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human vWF-A2 (aa 1-2813; NP_000543.2). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction