Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWF Monoclonal Antibody (907CT12.1.9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537656
Description
VWF Monoclonal Antibody for Western Blot, IHC (P), Flow
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.
Specifications
VWF | |
Monoclonal | |
0.48 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P04275 | |
VWF | |
VWF recombinant protein. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
907CT12.1.9 | |
Unconjugated | |
VWF | |
6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312 | |
Mouse | |
Protein G | |
RUO | |
7450 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction