Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWF Mouse anti-Human, Clone: 21-43, Abnova™

Catalog No. 89935102 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
125 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-102 125 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-102 Supplier Abnova Corporation Supplier No. MAB12551

Mouse monoclonal antibody raised against human VWF.

The glycoprotein encoded by this gene functions as both an antihemophilic factor carrier and a platelet-vessel wall mediator in the blood coagulation system. It is crucial to the hemostasis process. Mutations in this gene or deficiencies in this protein result in von Willebrand's disease. An unprocessed pseudogene has been found on chromosome 22. [provided by RefSeq]

Specifications

Antigen VWF
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 21-43
Conjugate Unconjugated
Formulation In PBS, pH 7.4 (50% glycerol, 0.2% BSA, 0.09% sodium azide).
Gene Alias F8VWF, VWD
Gene Symbols VWF
Host Species Mouse
Immunogen Human VWF
Purification Method Affinity Purified
Quantity 125 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage Store at 4°C. For long term storage store at −20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.