Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWF Mouse anti-Human, Clone: IIIE2.34, Abnova™

Catalog No. 89034496 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-496 Supplier Abnova Corporation Supplier No. MAB14413
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human VWF.

  • High specificity
  • High reactivity

Specifications

Antigen VWF
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone IIIE2.34
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human VWF.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Immunoprecipitation (0.5-1 ug/500 ug protein lysate) Western Blot (05-1 ug/mL
Formulation In PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P04275
Gene Alias F8VWF/VWD
Gene Symbols VWF
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 845-949 of human VWM.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.