Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWF Mouse anti-Human, Clone: OTI2H9, liquid, TrueMAB™
SDP

Catalog No. 501679785
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9785 Supplier OriGene Supplier No. TA803497
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor VIII related antigen or von Willebrand factor is a multimeric glycoprotein, which binds to platelet glycoprotein Ib, glycoprotein IIb/IIIa, collagen and heparin. It is synthesized by endothelial cells and stored in the Weibel-Palade granules. It mediates platelet adhesion to injured vessel walls and serves as a carrier and stabilizer for coagulation factor VIII. von Willebrand factor is one of the most useful markers to identify endothelial (or megakaryocytic) lineage of neoplasms. As not all endothelial cells synthesize/ store) this molecule, about 30% of tumors of vascular origin fail to stain for factor VIII related antigen, regardless of whether they are benign or maligt. Staining for factor VIII related antigen has also been used to measure angiogenesis, an indicator of tumor recurrence.

Specifications

Antigen VWF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene VWF
Gene Accession No. P04275
Gene Alias F8VWF, VWD
Gene Symbols VWF
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 764-1263 of human VWF produced in SF9 cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.