Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWF Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986153
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6153 Supplier Bioss Supplier No. BS4754R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.

Specifications

Antigen VWF
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VWF
Gene Accession No. P04275
Gene Alias 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand factor; Von Willebrand factor homolog; VWD; VWF; vwf {ECO:0000312
Gene Symbols VWF
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VWF/von Willebrand antigen 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.