Learn More
VWF/VWFpp Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 117781AP150UL

Description
This antibody raised against the N-terminal region of pre-pro-vWF recognizes von Willebrand antigen 2 (75-100 kDa) and pro-vWF (309-320 kDa).
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.Specifications
VWF/VWFpp | |
Polyclonal | |
Unconjugated | |
VWF | |
F8VWF, von Willebrand antigen II, von Willebrand factor, VWD, VWF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116669, 22371, 7450 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04275, Q62935, Q8CIZ8 | |
VWF | |
VWF, VWFpp Fusion Protein Ag2386 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.