Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VXN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$336.16 - $691.20
Specifications
| Antigen | C8orf46 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
VXN Polyclonal specifically detects VXN in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| C8orf46 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 254778 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QGAEASLPLTGSASCGVPGILRKMWTRHKKKSEYVGATNSAFEA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| chromosome 8 open reading frame 46, hypothetical protein LOC254778, MGC33510 | |
| C8ORF46 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title