Learn More
Invitrogen™ Vybrant™ DyeCycle™ Violet Ready Flow™ Reagent
Description
Vybrant DyeCycle Violet Ready Flow Reagent is a bright, easy-to-use, double-stranded DNA stain for cell cycle analysis in living cells using flow cytometry. It is excitable by a 405-nm laser with an emission peak at 437 nm.
• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh, or pipette
• Accurate cell-cycle analysis in living cells
• Low toxicity for cell sorting and additional live cell experiments
Vybrant DyeCycle Violet Ready Flow Reagent is a ready-to-use, direct-to-sample DNA stain. Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze.
As a cell-permeable dye, Vybrant DyeCycle Violet Ready Flow enters live cells and selectively binds dsDNA and emits a signal proportional to the DNA mass. The dye can be detected when excited with a 405-nm laser, and the emission captured at a peak of 437 nm. Vybrant DyeCycle Violet reagent is an ideal choice for DNA content analysis and can be included in multi-color cell cycle studies.
Specifications
Specifications
| Color | Violet |
| Content And Storage | 2 dropper bottles, store at room temperature |
| Product Type | Ready Flow Reagent |
| Product Line | Ready Flow, Vybrant |
| Quantity | 120 reactions |
| Shipping Condition | Room Temperature |
| Sub Cellular Localization | Nucleus |
| Emission Wavelength | 369/437 |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.