Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Potent and selective VHL-based dual Degrader (PROTAC™) of BRD7/9 (DC50 values are 4 and 34 nM for BRD7 and BRD9, respectively). Exhibits selectivity for BRD7/9 degradation over other bromodomain-containing proteins and other BAF/PBAF subunits. Displays cytotoxicity towards EOL-1 and A-204 cancer cell lines.
Negative control cis VZ 185 (Cat. No. 6939) also available.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2S,4R)-N-(2-((5-(4-(2,6-Dimethoxy-4-(2-methyl-1-oxo-1,2-dihydro-2,7-naphthyridin-4-yl)benzyl)piperazin-1-yl)pentyl)oxy)-4-(4-methylthiazol-5-yl)benzyl)-1-((S)-2-(1-fluorocyclopropane-1-carboxamido)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxamide |
| CAS | 2306193-61-1 |
| Quantity | 5 mg |
| Molecular Formula | C53H67FN8O8S |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

