W-13 hydrochloride, Tocris

Calmodulin antagonist

Manufacturer: R&D Systems 0361/50

Quantity 50mg

 View more versions of this product

Catalog No. 03-615-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C14H17ClN2O2S·HCl
    Formula Weight 349.27
    Synonym N-(4-Aminobutyl)-5-chloro-2-naphthalenesulfonamide hydrochloride
    Chemical Name or Material W-13 hydrochloride

    Specifications

    Quantity 50mg
    Formula Weight 349.27
    Assay Percent Range >98%
    Molecular Formula C14H17ClN2O2S·HCl
    CAS 88519-57-7
    Synonym N-(4-Aminobutyl)-5-chloro-2-naphthalenesulfonamide hydrochloride
    Recommended Storage Store at Room Temperature
    Chemical Name or Material W-13 hydrochloride

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit