Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ W146
GREENER_CHOICESDP

Catalog No. 36021R Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-021-R 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 36-021-R Supplier R&D Systems™ Supplier No. 3602/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective S1P1 receptor antagonist

W146 is a potent sphingosine-1-phosphate receptor S1P1 selective antagonist (Ki = 18 nM); displays no effect at S1P2, S1P3 or S1P5. Enhances capillary leakage and restores lymphocyte egress in vivo.

Specifications

Quantity 1 mg
Product Line Tocris
Product Type W146
Purity >98%
Molecular Weight (g/mol) 342.37
Inhibitors W146

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.