Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WAC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557705
Description
Immunogen sequence: RLSDGCHDRR GDSQPYQALK YSSKSHPSSG DHRHEKMRDA GDPSPPNKML RRSDSPENKY SDSTGHSKAK NVHTHRVRER DGGTSYSPQE NSH Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 91%.
The WW domain containing adaptor with coiled-coil protein (WAC) contains a WW domain that mediates protein-protein interactions and colocalizes with RNA splicing factor SC35. Further studies have indicated that WAC is a functional partner of the RNF20/40 complex that ubiquitinates Histone H2B, and that WAC regulates H2B ubiquitination. WAC targets RNF20/40 to associate with RNA polymerase II complex for H2B ubiquitination at active transcription sites. WAC-dependent transcription is also important for cell-cycle checkpoint activation in response to genotoxic stress.
Specifications
| WAC | |
| Polyclonal | |
| Unconjugated | |
| Wac | |
| 1110067P07Rik; A230035H12Rik; AI256735; AI256776; BM-016; DESSH; Kiaa1844; PRO1741; RGD1562407; Wac; WW domain containing adaptor with coiled-coil; WW domain-containing adapter protein with coiled-coil; WW domain-containing adaptor with coiled-coil; WW domain-containing protein 4; Wwp4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51322 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BTA9 | |
| Wac | |
| Recombinant protein corresponding to Human WAC. Recombinant protein control fragment (Product #RP-97019). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction