Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WACA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116364
Description
WACA Polyclonal Antibody for Western Blot
Acts as a linker between gene transcription and histone H2B monoubiquitination at 'Lys-120' (H2BK120ub1). Positive regulator of amino acid starvation-induced autophagy. Positively regulates MTOR activity. May negatively regulate the ubiquitin proteasome pathway. [UniProt].
Specifications
WACA | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q7ZUK7 | |
waca | |
KLH conjugated synthetic peptide between 487-519 amino acids from DANRE waca. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
waca | |
wac; waca; wu:fc13f04; wu:fj68f01; WW domain containing adaptor with coiled-coil; WW domain containing adaptor with coiled-coil a; WW domain-containing adapter protein with coiled-coil; WW domain-containing adapter with a coiled-coil region; zgc:55755 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
323904 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction