Learn More
Invitrogen™ WACA Polyclonal Antibody

Description
WACA Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | WACA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.77 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | waca |
| Gene Accession No. | Q7ZUK7 |
| Gene Alias | wac; waca; wu:fc13f04; wu:fj68f01; WW domain containing adaptor with coiled-coil; WW domain containing adaptor with coiled-coil a; WW domain-containing adapter protein with coiled-coil; WW domain-containing adapter with a coiled-coil region; zgc:55755 |
| Gene Symbols | waca |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.