Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Urinary System Model
SCI_ED

Catalog No. s07149
Click to view available options
:
Dimensions: 7.5 x 5 x 13.5 in. (19 x 13 x 34cm)

Free standing model representing major components of urinary system.

  • Comes with four parts
  • Right kidney is dissected to show cortex, medulla, pyramids, calyces, pelvis and origins of renal artery and vein
  • Bladder can also be removed and opened for detailed learning
  • Durable and great learning tool in all classrooms and laboratories
  • Mounted on a 10.5 x 7.5 in. board
  • Dimesnsions: 7.5 x 5 x 13.5 in. (19 x 13 x 34cm)

Specifications

For Use With (Application) For classroom and laboratory use
Includes Cortex, medulla, pyramids, calyces, pelvis and origins of the renal artery and vein
Product Type Model, Urinary System
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.