Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alere Clearview™ hCG Rapid Lateral Flow Test Kits

Catalog No. p-2836365
Click to view available options
Clearview Hcg Controls
Clearview Hcg Ii Urine Pk/50
Clrview Hcg Combo Ref Ln 30/pk
This item is not returnable. View return policy
This item is not returnable. View return policy

Results in five minutes or less

•    CLIA-waived for urine; non-waived for serum
•    Simple, one-step procedure
•    Tests for urine or serum
•    Results in five minutes or less
•    Built-in controls
•    Sensitivity: 20mIU/mL for urine; 10mIU/mL for serum
•    Specificity: 100%
•    Room temperature storage with up to 24 months dating

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.