Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WAPL Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100552H
Description
WAPL Polyclonal antibody specifically detects WAPL in Human samples. It is validated for Western BlotSpecifications
| WAPL | |
| Polyclonal | |
| Western Blot | |
| FOEKIAA0261friend of EBNA2 (Epstein-Barr virus nuclear protein 2), Friend of EBNA2 protein, WAPLwings apart-like protein homolog, wings apart-like homolog (Drosophila) | |
| The immunogen recognized by this antibody maps to a region between residues 1125 and 1175 of human wings apart like/KIAA0261 using the numbering given in TrEMBL entry Q7Z5K2 (GeneID 23063). | |
| 0.1 mL | |
| DNA Repair | |
| 23063 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction