Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WAPL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100552
Description
WAPL Polyclonal specifically detects WAPL in Human samples. It is validated for Western Blot.Specifications
WAPL | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
FOEKIAA0261friend of EBNA2 (Epstein-Barr virus nuclear protein 2), Friend of EBNA2 protein, WAPLwings apart-like protein homolog, wings apart-like homolog (Drosophila) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
NB100-552 is specific for human WAPL protein. | |
Store at 4C. Do not freeze. |
Western Blot | |
1.0 mg/mL | |
Western Blot 1:2500-1:10000 | |
Q7Z5K2 | |
WAPAL | |
The immunogen recognized by this antibody maps to a region between residues 1125 and 1175 of human wings apart like/KIAA0261 using the numbering given in TrEMBL entry Q7Z5K2 (GeneID 23063). | |
100 μL | |
DNA Repair | |
23063 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction