Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WAPL Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987968
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7968 Supplier Bioss Supplier No. BS7733R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcriptional repressor which may play a role in development of the central nervous system (CNS).

Specifications

Antigen WAPL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene WAPL
Gene Accession No. Q7Z5K2
Gene Alias A530089A20Rik; BC037674; DIF-2; Dioxin-inducible factor 2; FOE; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); Friend of EBNA2 protein; KIAA0261; RGD1308960; WAPAL; Wapl; WAPL cohesin release factor; wings apart-like homolog (Drosophila); Wings apart-like protein homolog
Gene Symbols WAPL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human WAPL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23063
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.