Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WAPL Recombinant Rabbit Monoclonal Antibody (3Q4G1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538145
Description
Positive Samples: HeLa, RD, Jurkat, C2C12 Immunogen sequence: MTSRFGKTYS RKGGNGSSKF DEVFSNKRTT LSTKWGETTF MAKLGQKRPN FKPDIQEIPK KPKVEEESTG DPFGFDSDDE SLPVSSKNLA QVKCSSYSES.
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
Specifications
WAPL | |
Recombinant Monoclonal | |
0.37 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q65Z40, Q7Z5K2 | |
WAPL | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human WAPL (Q7Z5K2). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
3Q4G1 | |
Unconjugated | |
WAPL | |
A530089A20Rik; BC037674; DIF-2; Dioxin-inducible factor 2; FOE; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); friend of EBNA2 protein; KIAA0261; RGD1308960; WAPAL; WAPL; WAPL cohesin release factor; wings apart-like homolog (Drosophila); wings apart-like protein homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
218914, 23063 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction