Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576817
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576817 Supplier Thermo Scientific Supplier No. PA576817
Add to Cart
Add to Cart

WARS Polyclonal Antibody

WARS Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen WARS
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene WARS1
Gene Accession No. P23381, P32921, Q6P7B0
Gene Alias GAMMA-2; hWRS; IFI 53; IFI53; IFP 53; IFP53; Interferon-induced protein 53; T1-TrpRS; T2-TrpRS; TrpRS; tryptophan tRNA ligase 1, cytoplasmic; tryptophan--tRNA ligase, cytoplasmic; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, cytoplasmic; Wars; WARS protein; Wrs
Gene Symbols WARS1
Host Species Rabbit
Immunogen Recombinant full length Human WARS.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22375, 314442, 7453
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.