Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100806
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100806 Supplier Invitrogen™ Supplier No. PA5100806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TrpRS.

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WARS2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WARS2
Gene Accession No. Q9CYK1, Q9UGM6
Gene Alias (Mt)TrpRS; 5730427B17Rik; 9430020O07Rik; AI413375; mitochondrial rryptophanyl-tRNA synthetase-like protein; mitochondrial tryptophanyl-tRNA synthetase 2; trpRS; tryptophan tRNA ligase 2, mitochondrial; tryptophan--tRNA ligase, mitochondrial; tryptophanyl tRNA synthetase 2 (mitochondrial); tryptophanyl tRNA synthetase 2, mitochondrial; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, mitochondrial; tryptophanyl-tRNA synthetase, mitochondrial; tryptophan--tRNA ligase, mitochondrial; WARS2; zgc:110828
Gene Symbols WARS2
Host Species Rabbit
Immunogen A synthesized peptide derived from human WARS2(Accession Q9UGM6), corresponding to amino acid residues K199-Q249.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10352, 70560
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.