Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567064 Supplier Invitrogen™ Supplier No. PA567064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MELVQDLAQGF NKKYGEFFPV PESILTSMKK VKSLRDPSAK MSKSDPDKLA TVRITDSPEE IVQKFRKAVT DFTSEVTYDP A Highest antigen sequence identity to the following orthologs - mouse 90%, rat 88%.

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WARS2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WARS2
Gene Accession No. Q9UGM6
Gene Alias (Mt)TrpRS; 5730427B17Rik; 9430020O07Rik; AI413375; mitochondrial rryptophanyl-tRNA synthetase-like protein; mitochondrial tryptophanyl-tRNA synthetase 2; trpRS; tryptophan tRNA ligase 2, mitochondrial; tryptophan--tRNA ligase, mitochondrial; tryptophanyl tRNA synthetase 2 (mitochondrial); tryptophanyl tRNA synthetase 2, mitochondrial; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, mitochondrial; tryptophanyl-tRNA synthetase, mitochondrial; tryptophan--tRNA ligase, mitochondrial; WARS2; zgc:110828
Gene Symbols WARS2
Host Species Rabbit
Immunogen Recombinant Human WARS2. Recombinant protein control fragment (Product #RP-107704).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10352
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.