Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WASH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551731 Supplier Invitrogen™ Supplier No. PA551731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAPREVVDPS GGWATLLESI RQAGGIGKAK LRSMKERKLE KKKQKEQEQV RATSQGGHLM SDLFNKLVMR HKGISGKGPG AGEGPGGAFA RVSDSIPPLP PPQQPQVDED EDDWES Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

WASHC1 (WASH1) is a protein coding gene with endocytosis as a related pathway. Acts as a nucleation-promoting factor (NPF) at the surface of endosomes, where it recruits and activates the Arp2/3 complex to induce actin polymerization, playing a key role in the fission of tubules that serve as transport intermediates during endosome sorting. Its assembly in the WASH core complex seems to inhibit its NPF activity and via WASHC2 is required for its membrane targeting. Involved in endocytic trafficking of EGF. Involved in transferrin receptor recycling. Regulates the trafficking of endosomal alpha5beta1 integrin to the plasma membrane and involved in invasive cell migration. In T-cells involved in endosome-to-membrane recycling of receptors including T-cell receptor (TCR), CD28 and ITGAL; proposed to be implicated in T cell proliferation and effector function. In dendritic cells involved in endosome-to-membrane recycling of major histocompatibility complex (MHC) class II probably involving retromer and subsequently allowing antigen sampling, loading and presentation during T-cell activation. Involved in Arp2/3 complex-dependent actin assembly driving Salmonella typhimurium invasion independent of ruffling. Involved in the exocytosis of MMP14 leading to matrix remodeling during invasive migration and implicating late endosome-to-plasma membrane tubular connections and cooperation with the exocyst complex. Involved in negative regulation of autophagy independently from its role in endosomal sorting by inhibiting BECN1 ubiquitination to inactivate PIK3C3/Vps34 activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WASH1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WASHC1
Gene Accession No. A8K0Z3
Gene Alias CXYorf1-like protein on chromosome 9; FAM39E; family with sequence similarity 39, member E; FLJ00075 protein; Protein FAM39E; WAS protein family homolog 1; WASH; WASH complex subunit 1; WASH1; WASHC1
Gene Symbols WASHC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WASH1. Recombinant protein control fragment (Product #RP-92149).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100287171
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.