Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WASHC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | WASH4P |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
WASHC1 Polyclonal specifically detects WASHC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
WASH4P | |
Polyclonal | |
Rabbit | |
Human | |
A8MWX3 | |
374677 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GAPREVVDPSGGWATLLESIRQAGGIGKAKLRSMKERKLEKKKQKEQEQVRATSQGGHLMSDLFNKLVMRHKGISGKGPGAGEGPGGAFARVSDSIPPLPPPQQPQVDEDEDDWES | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CXYorf1P, FAM39CP, Family With Sequence Similarity 39, Member C Pseudogene, Protein FAM39CP, Pseudogene Of CXYorf1, Putative WAS Protein Family Homolog 4, WAS Protein Family Homolog 4 Pseudogene | |
WASH4P | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title