Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WASP Antibody (SR2069), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322175100UL
Description
WASP Monoclonal antibody specifically detects WASP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
WASP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR2069 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:40 | |
IMD2THC, WASP, WiskTHC1, wiskthrombocytopenia 1 (X-linked) | |
A synthesized peptide derived from human WASP (Uniprot #: P42768) | |
100 μg | |
Phospho Specific, Signal Transduction | |
7454 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction