Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WASP, p Ser483, p Ser484 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002307V
Description
WASP Polyclonal antibody specifically detects WASP in Human samples. It is validated for Western BlotSpecifications
WASP | |
Polyclonal | |
Western Blot | |
IMD2THC, WASP, WiskTHC1, wiskthrombocytopenia 1 (X-linked) | |
The immunogen this antibody was made to, is phosphorylated Serine 483, phosphorylated Serine 484 and surrounding residues of human Wiskott-Aldrich syndrome using the numbering given in entry NP_000368.1 (GeneID 7454). | |
0.1 mL | |
Phospho Specific, Signal Transduction | |
7454 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction